Learn More
*AGAROSE BEAD TECHNOLOGIES Low Den. Glyoxal 4% Rapid Run™-25mL
Catalog No. NC1755562
Encompass_Preferred
Low Density Glyoxal 4 Rapid Run™ allows a covalent binding of agarose to lysine amino groups. This product is adequate to work in large scale process (High Pressure). The resin has a low content of aldehydes groups (15- 25 µmoles Glyoxal /ml gel) providing a minimum distortion of conjugated affinity ligand. The resin has been manufactured using 4% Rapid Run™ agarose beads, because of this, the pore size is adequate to work with enzymes, proteins & antibodies and the stability at high flow is adequate to large scale process.Tech Specs:Description : High Flow Minimum distortion of immobilized ligand. Exclusion limit: ~3×10^7Activation degree (μmol glyoxyl/ml gel) : 15-25
Catalog No. NC1755562 Supplier *AGAROSE BEAD TECHNOLOGIES Supplier No. 4RRGL025
Low Density Glyoxal 4 Rapid Run™ allows a covalent binding of agarose to lysine amino groups. This product is adequate to work in large scale process (High Pressure). The resin has a low content of aldehydes groups (15- 25 µmoles Glyoxal /ml gel) providing a minimum distortion of conjugated affinity ligand. The resin has been manufactured using 4% Rapid Run™ agarose beads, because of this, the pore size is adequate to work with enzymes, proteins & antibodies and the stability at high flow is adequate to large scale process.Tech Specs:Description : High Flow Minimum distortion of immobilized ligand. Exclusion limit: ~3×10^7Activation degree (μmol glyoxyl/ml gel) : 15-25