Learn More
Kalen Biomedical LLC LOW DENSITY LIPOPROTE LDL 5MG
Catalog No. NC9835064
Encompass_Preferred
Human Low Density Lipoprotein (d=1.019 - 1.063 g/mL) is isolated from plasma. The product is provided in PBS with EDTA as a preservative. The protein concentration is 5 mg/mL and the endotoxin level is less than 0.5 EU /mg of protein.
Catalog No. NC9835064 Supplier Kalen Biomedical LLC Supplier No. 7702004
Human Low Density Lipoprotein (d=1.019 - 1.063 g/mL) is isolated from plasma. The product is provided in PBS with EDTA as a preservative. The protein concentration is 5 mg/mL and the endotoxin level is less than 0.5 EU /mg of protein.