Learn More
Cusabio Technology LLC low density lipoprotein LDL
Catalog No. 5000051348
Encompass_Preferred
This kit detects LDL (low-density lipoprotein) in rat serum plasma and tissue homogenates using a competitive ELISA method It provides a quantitative measurement with a detection range of 0 312 g/mL to 20 g/mL and a sensitivity of 0 156 g/mL ensuring precise analysis of LDL levels The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Readings are taken at an absorbance wavelength of 450 nm offering reliable results for research applications in cardiovascular and metabolic studies The kit is designed for efficiency and accuracy supporting standardized protocols to streamline experimental workflows while maintaining reproducibility across biological samples
Catalog No. 50-000-51348 Supplier Cusabio Technology LLC Supplier No. CSBE11704R
This kit detects LDL (low-density lipoprotein) in rat serum plasma and tissue homogenates using a competitive ELISA method It provides a quantitative measurement with a detection range of 0 312 g/mL to 20 g/mL and a sensitivity of 0 156 g/mL ensuring precise analysis of LDL levels The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Readings are taken at an absorbance wavelength of 450 nm offering reliable results for research applications in cardiovascular and metabolic studies The kit is designed for efficiency and accuracy supporting standardized protocols to streamline experimental workflows while maintaining reproducibility across biological samples