Learn More
Waters Corp Waters Corporation Tubing Assembly, PEEK, 0.005 in. ID, 20 in. L, Red
Catalog No. 50791881 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This low dispersion red line tubing assembly is designed for high-performance liquid chromatography (HPLC) systems. It features PEEK construction with a 0.005 in. internal diameter and a 20 in. length, equipped with one piece fingertight fittings for secure connections.
  • Tubing for HPLC systems.
  • Constructed from PEEK material.
  • 0.005 in. internal diameter.
  • 20 in. length.
  • Equipped with one piece fingertight fittings.
  • Suitable for 2695 Separations Module and 2695 D Dissolution Separations Module.
  • Designed for HPLC systems.
  • Replacement as required.
Catalog No. 50-791-881 Supplier Waters Corp Supplier No. 430000922
This low dispersion red line tubing assembly is designed for high-performance liquid chromatography (HPLC) systems. It features PEEK construction with a 0.005 in. internal diameter and a 20 in. length, equipped with one piece fingertight fittings for secure connections.
  • Tubing for HPLC systems.
  • Constructed from PEEK material.
  • 0.005 in. internal diameter.
  • 20 in. length.
  • Equipped with one piece fingertight fittings.
  • Suitable for 2695 Separations Module and 2695 D Dissolution Separations Module.
  • Designed for HPLC systems.
  • Replacement as required.