Learn More
Sarstedt Inc SafeSeal reaction tube, 2 ml, PP, PCR Performance Tested, Low DNA-binding, 200/pack
Catalog No. NC0964896 Shop All Sarstedt Inc. Products
Encompass_Preferred
2mL SafeSeal reaction tube DNA low binding are certified PCR Performance Tested. There tubes come with attached lid with retaining cames and feature moulded graduation and frosted writing space.
Catalog No. NC0964896 Supplier Sarstedt Inc Supplier No. 72.695.700
2mL SafeSeal reaction tube DNA low binding are certified PCR Performance Tested. There tubes come with attached lid with retaining cames and feature moulded graduation and frosted writing space.