Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Low EDTA TE (1X), pH 8.0, Pack 4 X 100 mL
SDP

Catalog No. NC1247676
Encompass_Preferred

Low-EDTA TE (1X), pH 8.0 is molecular biology grade TE buffer used to store DNA and RNA. EDTA chelates Mg+2 and other divalent metals ions (inhibits DNAse and RNAse to suppress DNA and RNA degradation). Tris is a buffering agent to keep the solution at a defined pH The Cl or HCl indicates the pH was attained using aqueous hydrochloric acid (HCl), since Tris is naturally basic. Tris-EDTA (TE) is routinely used for suspending nucleic acid samples.

Contains 0.1mM EDTA

Applications: Low EDTA is recommended to re-suspend final purified DNA when DNA is used for downstream PCR

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
pH: 8.0 ± 0.1
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. NC1247676 Supplier Quality Biological Inc Supplier No. 351324721
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Low-EDTA TE (1X), pH 8.0 is molecular biology grade TE buffer used to store DNA and RNA. EDTA chelates Mg+2 and other divalent metals ions (inhibits DNAse and RNAse to suppress DNA and RNA degradation). Tris is a buffering agent to keep the solution at a defined pH The Cl or HCl indicates the pH was attained using aqueous hydrochloric acid (HCl), since Tris is naturally basic. Tris-EDTA (TE) is routinely used for suspending nucleic acid samples.

Contains 0.1mM EDTA

Applications: Low EDTA is recommended to re-suspend final purified DNA when DNA is used for downstream PCR

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
pH: 8.0 ± 0.1
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.