Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Low endotoxin gelatin from porcine skin

Catalog No. 9017565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-175-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-175-65G Supplier Sigma Aldrich Supplier No. 9017565G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gelatin has received significant attention in the biomedical field due to its inherent bioactivity. Low endotoxin gelatin from porcine skin, gel strength 300 bloom is a high bloom (high molecular weight) gelatin featuring less than 10 endotoxin units (EU) per gram. Endotoxins primarily consist of lipopolysaccharides from the outer membrane of gram-negative bacteria. Once in the bloodstream, endotoxins are capable of eliciting a strong immune response, potentially leading to fever, shock, a drop in blood pressure, and even potentially death. In addition to this immune response, several reports suggest endotoxins may also damage various types of cells. To avoid adverse effects, raw materials should contain the lowest level of endotoxins possible when used in biomedical or regenerative medicine research applications. This low endotoxin gelatin is derived from alkaline-treated porcine skin and has a variety of potential applications in the fields of regenerative medicine, medical devices, and tissue engineering research.beMatrix is a trademark of Nitta Gelatin

Specifications

CAS 9000-70-8
Quantity 5 g
Synonym beMatrix(TM) gelatin LS-H
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.