Learn More
Thomson Instrument Company Low Evap | Filter Vial, PTFE 0.2µm, Non-Slit Septum, Green Screw Cap
SDP
Catalog No. NC1792990
Encompass_Preferred
Low Evap|Filter Vial are available in all three membranes types; PTFE, PVDF, & Nylon and two pore sizes 0.2μm & 0.45μm and also feature our color coded system of caps to help selection in the lab.Low Evap|Filter Vial are suitable for GC/GCMS Applications, Solubility Testing, Food Testing, Volatiles or any other application where minimizing the evaporation of samples is critical.
• Eliminate the need for syringe filters, syringes, HPLC vials and caps
• Clear up valuable lab space
• Offer Eco-Friendly solution to lowering your lab waste
• Evaporation rate <0.4% over 24-hour
• Made in the USA
Catalog No. NC1792990 Supplier Thomson Instrument Company Supplier No. 64430
Low Evap|Filter Vial are available in all three membranes types; PTFE, PVDF, & Nylon and two pore sizes 0.2μm & 0.45μm and also feature our color coded system of caps to help selection in the lab.Low Evap|Filter Vial are suitable for GC/GCMS Applications, Solubility Testing, Food Testing, Volatiles or any other application where minimizing the evaporation of samples is critical.
• Eliminate the need for syringe filters, syringes, HPLC vials and caps
• Clear up valuable lab space
• Offer Eco-Friendly solution to lowering your lab waste
• Evaporation rate <0.4% over 24-hour
• Made in the USA