Learn More
Waters Corp Waters Nebulizer, Low Flow, 50 to 500 μL/min
Catalog No. 50791684 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Low Flow Nebulizer is a service part designed to provide precise flow control for analytical and microbore applications. It is an optional component, compatible with the 2424 Evaporative Light Scattering (ELS) Detector.
  • Provides a flow rate of 50 to 500 μL/min.
  • Requires a gas flow of 0.77 L/min at 25 psi.
  • Functions as a spare parts kit for maintenance and replacement.
Catalog No. 50-791-684 Supplier Waters Corp Supplier No. 205000459
The Low Flow Nebulizer is a service part designed to provide precise flow control for analytical and microbore applications. It is an optional component, compatible with the 2424 Evaporative Light Scattering (ELS) Detector.
  • Provides a flow rate of 50 to 500 μL/min.
  • Requires a gas flow of 0.77 L/min at 25 psi.
  • Functions as a spare parts kit for maintenance and replacement.