Learn More
Zeflon International, Inc Low Flow Rotameter Flowmeter, 0-4 LPM, Non-adjustable
SDP
Catalog No. NC0806062
Encompass_Preferred
Low Flow Rotameter Flowmeter, 0-4 LPM. Zefon Field Rotameters are a low cost way to measure airflow in air sampling applications. They feature a durable acrylic housing with direct reading scales printed on them. Available in three different flow ranges covering a range of 0.2 through 30 LPM. Operation and maintenance are very simple; only a few common sense precautions must be observed to assure long, trouble-free service.
Catalog No. NC0806062 Supplier Zeflon International, Inc Supplier No. 116116
Low Flow Rotameter Flowmeter, 0-4 LPM. Zefon Field Rotameters are a low cost way to measure airflow in air sampling applications. They feature a durable acrylic housing with direct reading scales printed on them. Available in three different flow ranges covering a range of 0.2 through 30 LPM. Operation and maintenance are very simple; only a few common sense precautions must be observed to assure long, trouble-free service.