Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Metalube and Coating Services Degreaser, Ph 13, 55 gal

Catalog No. 502820106
Encompass

This concentrated low foam cleaner degreaser is designed for aqueous cleaning applications. It is intended to be diluted prior to use and is effective in industrial cleaning environments.

  • Liquid physical state
  • Amber appearance
  • Mild odor
  • Ph of 13
  • Boiling point of 98.89 °C
  • Specific gravity of 1.07
  • Completely soluble in water

Catalog No. 50-282-0106 Supplier MSC Supplier No. 18726521
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This concentrated low foam cleaner degreaser is designed for aqueous cleaning applications. It is intended to be diluted prior to use and is effective in industrial cleaning environments.

  • Liquid physical state
  • Amber appearance
  • Mild odor
  • Ph of 13
  • Boiling point of 98.89 °C
  • Specific gravity of 1.07
  • Completely soluble in water

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.