Learn More
Cole-Parmer Cole-Parmer elements Low-Form Beaker, Glass, 100 mL, 12/pk
Catalog No. NC1470678 Shop All Cole-Parmer Products
Encompass_Preferred
These beakers are constructed of durable borosiicate 3.3 glass with pour spout which is ideal for general use at an economical price.
Catalog No. NC1470678 Supplier Cole-Parmer Supplier No. 3450240
These beakers are constructed of durable borosiicate 3.3 glass with pour spout which is ideal for general use at an economical price.