Learn More
Cole-Parmer Cole-Parmer elements Plus Griffin Low-Form Beaker, Glass, 5000 mL, 1/EA
Catalog No. NC1698383 Shop All Cole-Parmer Products
Encompass_Preferred
These low-form beakers are constructed of Class A borosilicate glass to meet ASTM E960 specifications.
Catalog No. NC1698383 Supplier Cole-Parmer Supplier No. 3450337
These low-form beakers are constructed of Class A borosilicate glass to meet ASTM E960 specifications.