Learn More
Chemglass Life Sciences Chemglass Pyrex Griffin Low-Form Beaker, 600 mL, Double Scale
SDP
Catalog No. NC3684544
Encompass_Preferred
This 600 mL low-form (Griffin) beaker is a general-purpose graduated vessel suitable for mixing, heating, and liquid handling in laboratory environments. It is made to Type I glass standards with uniform wall thickness for strength and thermal shock resistance.
  • 600 mL capacity.
  • Low-form (Griffin) design with pouring spout.
  • Double-scale graduations; 50 mL intervals from 50-500 mL.
  • Type I borosilicate glass for chemical and thermal resistance.
  • Uniform wall thickness for improved mechanical strength and thermal shock resistance.
  • Approx. OD x height: 90 x 124 mm.
  • Packaging may vary by supplier; manufacturer lists single units (each).
Catalog No. NC3684544 Supplier Chemglass Life Sciences Supplier No. CG8048600
This 600 mL low-form (Griffin) beaker is a general-purpose graduated vessel suitable for mixing, heating, and liquid handling in laboratory environments. It is made to Type I glass standards with uniform wall thickness for strength and thermal shock resistance.
  • 600 mL capacity.
  • Low-form (Griffin) design with pouring spout.
  • Double-scale graduations; 50 mL intervals from 50-500 mL.
  • Type I borosilicate glass for chemical and thermal resistance.
  • Uniform wall thickness for improved mechanical strength and thermal shock resistance.
  • Approx. OD x height: 90 x 124 mm.
  • Packaging may vary by supplier; manufacturer lists single units (each).