Learn More
Alkali Scientific Low Form Beaker with Handle, Printed Graduations, and Anti Slip Base, Polypropylene (PP), 500mL, 24/Case
SDP
Catalog No. 5000662080
Encompass_Preferred
The Low Form Polypropylene Beaker is designed for safe and accurate liquid handling in laboratory, research, and production environments. Molded from durable, chemically resistant polypropylene, this beaker provides excellent strength and clarity for routine mixing, sample preparation, and heating within polypropylene temperature limits. The wide, low form design improves stability on benches and hot plates, while the integrated ergonomic handle allows controlled one handed pouring. Bold printed graduations provide quick visual volume checks, helping improve measurement accuracy during routine lab work. A textured anti slip base enhances grip on smooth surfaces, reducing the risk of spills or tipping. Ideal for academic laboratories, biotechnology, pharmaceutical, industrial testing, and general laboratory workflows requiring reliable liquid handling.
Catalog No. 50-006-62080 Supplier Alkali Scientific Supplier No. LFB0500H
Add to Cart
Add to Cart
The Low Form Polypropylene Beaker is designed for safe and accurate liquid handling in laboratory, research, and production environments. Molded from durable, chemically resistant polypropylene, this beaker provides excellent strength and clarity for routine mixing, sample preparation, and heating within polypropylene temperature limits. The wide, low form design improves stability on benches and hot plates, while the integrated ergonomic handle allows controlled one handed pouring. Bold printed graduations provide quick visual volume checks, helping improve measurement accuracy during routine lab work. A textured anti slip base enhances grip on smooth surfaces, reducing the risk of spills or tipping. Ideal for academic laboratories, biotechnology, pharmaceutical, industrial testing, and general laboratory workflows requiring reliable liquid handling.