Learn More
Alkali Scientific Low Form Beaker with Printed Graduations, and Anti Slip Base, Stackable, Polypropylene, 100mL, 48/Case
SDP
Catalog No. 5000662084
Encompass_Preferred
These low form Griffin style beakers are engineered for reliable performance in routine laboratory mixing, pouring, stirring, and solution preparation. Molded from virgin polypropylene (PP), they provide a safe, shatterproof alternative to glass while offering excellent value, durability, and chemical resistance. Precision automated molding ensures consistent quality, and the single non drip spout delivers clean, controlled pouring. For convenient storage, all handle less models feature a fully stackable design. Low form Griffin design for versatile laboratory use Manufactured from virgin, USP Class VI polypropylene (PP) Meets FDA 21 CFR and European Pharmacopeia requirements Bold, printed dual scale graduations for easy volume reading Single non drip spout for accurate, spill free pouring Autoclavable at 121°C Resistant to most acids, bases, and many common solvents Stackable, space saving design (no handle version)
Catalog No. 50-006-62084 Supplier Alkali Scientific Supplier No. LFB0100
Add to Cart
Add to Cart
These low form Griffin style beakers are engineered for reliable performance in routine laboratory mixing, pouring, stirring, and solution preparation. Molded from virgin polypropylene (PP), they provide a safe, shatterproof alternative to glass while offering excellent value, durability, and chemical resistance. Precision automated molding ensures consistent quality, and the single non drip spout delivers clean, controlled pouring. For convenient storage, all handle less models feature a fully stackable design. Low form Griffin design for versatile laboratory use Manufactured from virgin, USP Class VI polypropylene (PP) Meets FDA 21 CFR and European Pharmacopeia requirements Bold, printed dual scale graduations for easy volume reading Single non drip spout for accurate, spill free pouring Autoclavable at 121°C Resistant to most acids, bases, and many common solvents Stackable, space saving design (no handle version)