Learn More
Andwin Scientific LOW FORM BEAKER W/HANDLE
SDP
Supplier:  Andwin Scientific 13916730
Encompass_Preferred
The Graduated Beaker with Handle are sturdy beakers made of autoclavable, translucent polypropylene. Its low form structure provides better stability. This product is graduated in the range of 501000 mL, which allows precise sample measurement.
Catalog No. NC9846530