Learn More
Qorpak™ Low-form Polypropylene Pitcher with Handle
Description
- Molded graduations will not wear off
- Large strong handles provides safer handling of acids, alkalis, and other hazardous solutions
Specifications
Specifications
| Color | Natural |
| Capacity (Metric) | 2 L |
| Material | Polypropylene |
| Graduation Range | 100 mL |
| Quantity | 6/Pk. |
| Type | Pitcher |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Scale | Molded Scale |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.