Learn More
Enzo Life Sciences BIOARRAY™ Low Input RNA Amplification and Biotin Labeling System (10Reactions)
SDP
Catalog No. 502012517
Encompass_Preferred
The BIOARRAY™ Low Input RNA Amplification with Biotin Labeling System provides an optimized protocol and reagents for the production of biotin-labeled antisense RNA (aRNA) from total cellular RNA samples in less than 24 hours for array analysis. This kit has been improved to yield sufficient aRNA quantity for most microarray platforms (10 µg minimum) with total RNA input of as low as 20 ng and up to 500 ng. Entire amplification reaction can be performed in a single tube and the biotin-labeled aRNA can be purified using either magnetic beads or purification columns. The BIOARRAY™ Low Input aRNA Amplification and Biotin Labeling System has been optimized for superior performance with reduced variability and improved reproducibility and data quality. The complete system is composed of reagents for cDNA synthesis and in vitro transcript labeling.  
Catalog No. 50-201-2517 Supplier Enzo Life Sciences Supplier No. ENZ424220010
The BIOARRAY™ Low Input RNA Amplification with Biotin Labeling System provides an optimized protocol and reagents for the production of biotin-labeled antisense RNA (aRNA) from total cellular RNA samples in less than 24 hours for array analysis. This kit has been improved to yield sufficient aRNA quantity for most microarray platforms (10 µg minimum) with total RNA input of as low as 20 ng and up to 500 ng. Entire amplification reaction can be performed in a single tube and the biotin-labeled aRNA can be purified using either magnetic beads or purification columns. The BIOARRAY™ Low Input aRNA Amplification and Biotin Labeling System has been optimized for superior performance with reduced variability and improved reproducibility and data quality. The complete system is composed of reagents for cDNA synthesis and in vitro transcript labeling.  