Learn More
Electron Microscopy Sciences Low-Intensity UV Lamp, 110 V
SDP
Catalog No. 50949647
Encompass_Preferred
This 6-watt UV lamp meets most laboratory requirements; such as, bacterial identification, fluorchemistry, specimen staining, titration processes, photochemical reactions, aflatoxin detection, and curing UV cements. Constructed from Cyolac plastic, with a longwave emission of 365nm and an intensity at 6" (15.2cm) of 750 um/cm squared. The bulb has a rated life of 6,000 hours. It can be used as a hand-held light or it may br mounted on a optional stand.
Catalog No. 50-949-647 Supplier Electron Microscopy Sciences Supplier No. 72422
This 6-watt UV lamp meets most laboratory requirements; such as, bacterial identification, fluorchemistry, specimen staining, titration processes, photochemical reactions, aflatoxin detection, and curing UV cements. Constructed from Cyolac plastic, with a longwave emission of 365nm and an intensity at 6" (15.2cm) of 750 um/cm squared. The bulb has a rated life of 6,000 hours. It can be used as a hand-held light or it may br mounted on a optional stand.