Learn More
Hampton Research HR2-120/Low Ionic Strength Screen - 24 unique reagents of varying pH and PEG 3,350 concentrations, 1 ml each
SDP
Catalog No. 502723
Encompass
HR2-120/Low Ionic Strength Screen - 24 unique reagents of varying pH and PEG 3,350 concentrations, 1 ml each (NOTE: HR2-120 and Dehydrant (HR2-519) are sold separately. Both items are needed to perform the screen.). Please note items from this supplier are non-returnable.
Catalog No. 50-272-3 Supplier Hampton Research Supplier No. HR2120
HR2-120/Low Ionic Strength Screen - 24 unique reagents of varying pH and PEG 3,350 concentrations, 1 ml each (NOTE: HR2-120 and Dehydrant (HR2-519) are sold separately. Both items are needed to perform the screen.). Please note items from this supplier are non-returnable.