Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Low-Lint Nylon Women's Inspection Gloves

Catalog No. 19053703 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-053703 Small
19-053702 Medium
19-013620 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-053703
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Moisture-absorbent gloves are constructed of 40-denier nylon tricot fabric and can be worn alone, or used as a glove liner.

  • 40-denier tricot fabric in a two-piece construction
  • Easy size identification by color-coded hem on rolled cuff
  • Can be worn alone or used as a liner
  • Moisture absorbent

Specifications

Construction Cut and Sewn
Coating Material Uncoated
Color White
Disposable Limited-use
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Small
Sterility Non-sterile
For Use With (Application) Inspection, Electronics Assembly
Gender Women's
Glove Cuff Style Rolled
Hem Color Black
Series 98-741
Length (English) 8.5 in.
Length (Metric) 21 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.