Learn More
Meridian Life Science Low LOD 1-Step RT-qPCR Mix, 500ML
SDP
Catalog No. 5000856327
Encompass_Preferred
Low LOD 1-Step RT-qPCR Mix comprises a master mix containing Taq polymerase, reaction buffer, MgCl2 and optimized dNTP/dUTP mix, and separate reverse transcriptase/RNase inhibitor mix. Low LOD 1-Step RT-qPCR Mix is designed for stringent amplification of highly-structured and very low copy-number RNA and DNA targets, ideal for applications such as blood bank or transplant viral testing.
Catalog No. 50-008-56327 Supplier Meridian Life Science Supplier No. MDX025500
Low LOD 1-Step RT-qPCR Mix comprises a master mix containing Taq polymerase, reaction buffer, MgCl2 and optimized dNTP/dUTP mix, and separate reverse transcriptase/RNase inhibitor mix. Low LOD 1-Step RT-qPCR Mix is designed for stringent amplification of highly-structured and very low copy-number RNA and DNA targets, ideal for applications such as blood bank or transplant viral testing.