Learn More
Gold Biotechnology Inc Low Melt Agarose 100 g
SDP
Catalog No. NC2596266
Encompass
Low Melting (LM) Agarose is a low melting temperature agarose with the highest resolving capacity for large DNAfragments, including PCR products.
Catalog No. NC2596266 Supplier Gold Biotechnology Inc Supplier No. A204100
Low Melting (LM) Agarose is a low melting temperature agarose with the highest resolving capacity for large DNAfragments, including PCR products.