Learn More
Gold Biotechnology Inc Low Melt Agarose 25 g
SDP
Catalog No. 501532729
Encompass_Preferred
Low Melting (LM) Agarose is a low melting temperature agarose with the highest resolving capacity for large DNAfragments, including PCR products.
Catalog No. 50-153-2729 Supplier Gold Biotechnology Inc Supplier No. A20425
Low Melting (LM) Agarose is a low melting temperature agarose with the highest resolving capacity for large DNAfragments, including PCR products.