Learn More
New England Biolabs, Inc. Low Molecular Weight DNA Ladder – 0.5 ml
SDP
Catalog No. 50814110
Encompass
A proprietary plasmid is digested to completion with appropriate restriction enzymes to yield 11 bands suitable for use as molecular weight standards for both agarose and polyacrylamide gel electrophoresis. This digested DNA includes fragments ranging from 25-766 base pairs. The 200 base pair band has increased intensity to serve as a reference point. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Catalog No. 50-814-110 Supplier New England Biolabs, Inc. Supplier No. N3233L
A proprietary plasmid is digested to completion with appropriate restriction enzymes to yield 11 bands suitable for use as molecular weight standards for both agarose and polyacrylamide gel electrophoresis. This digested DNA includes fragments ranging from 25-766 base pairs. The 200 base pair band has increased intensity to serve as a reference point. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.