Learn More
Creative Biomart Urokinase
Catalog No. 502084072
Encompass_Preferred
The product, that consists of an A chain of 2,000 daltons linked by a sulfhydryl bond to a B chain of 30,400 daltons. Recombinant urokinase plasminogen activator.
Catalog No. 50-208-4072 Supplier Creative Biomart Supplier No. THP0021
The product, that consists of an A chain of 2,000 daltons linked by a sulfhydryl bond to a B chain of 30,400 daltons. Recombinant urokinase plasminogen activator.