Learn More
World Precision Instrument Low-noise cable for Microelectrode Holder, A: BNC (male), B: jack
SDP
Catalog No. 50822111
Encompass
For wiring any laboratory setup
This standard cable has a male BNC connector on one end and a 2mm gold jack on the other end. The cable is 4' (1.2m) long.
Catalog No. 50-822-111 Supplier World Precision Instrument Supplier No. 5375
For wiring any laboratory setup
This standard cable has a male BNC connector on one end and a 2mm gold jack on the other end. The cable is 4' (1.2m) long.