Learn More
Electron Microscopy Sciences Low-Odor Formalin 10% Buffered, 6.9 to 7.1 at 25°C pH 4 LT
SDP
Catalog No. 50980503
Encompass_Preferred
CH₂O
Low odor, prepared w/v, pH 6.9 to 7.1 at 25 degree C. 10% formalin in Phosphate buffer according to the A.F.I.P. Manual of Histology. This solution allows for long-term storage of tissue specimens.
pH: 6.9 to 7.1 at 25°C
Catalog No. 50-980-503 Supplier Electron Microscopy Sciences Supplier No. 1574101
CH₂O
Low odor, prepared w/v, pH 6.9 to 7.1 at 25 degree C. 10% formalin in Phosphate buffer according to the A.F.I.P. Manual of Histology. This solution allows for long-term storage of tissue specimens.
pH: 6.9 to 7.1 at 25°C