Learn More
AB Sciex LLC Low pH SDS Sample Buffer, 140mL
Catalog No. NC1784649
Encompass
Low pH SDS Sample Buffer, 140 mL (100 mM Tris-HCl, pH 6.8, 1% SDS) allows you to use SCIEX’s Gold Standard CE-SDS with a low pH SDS sample buffer. Use of this buffer can increase non-reduced purity by decreasing method induced fragmentation. Less variability and lower RSDs means less troubleshooting and faster method development.
Catalog No. NC1784649 Supplier AB Sciex LLC Supplier No. C44807
Low pH SDS Sample Buffer, 140 mL (100 mM Tris-HCl, pH 6.8, 1% SDS) allows you to use SCIEX’s Gold Standard CE-SDS with a low pH SDS sample buffer. Use of this buffer can increase non-reduced purity by decreasing method induced fragmentation. Less variability and lower RSDs means less troubleshooting and faster method development.