Learn More
Waters Corp Waters UPLC Method Development Kit, Low pH, 3 mm x 50 mm, 4 pk
Catalog No. 50791066 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method development kit is designed for low pH UPLC applications, incorporating advanced 1.7 μm BEH ShieldRP18 and Phenyl with 1.8 μm HSS C18 and C18 SB column chemistries. It facilitates various method development approaches, enabling efficient and effective analytical processes.
  • Accommodates diverse method development strategies.
  • Leverages UPLC technology for enhanced throughput and reduced analysis time.
  • Maintains separation integrity for reliable results.
  • Features reversed phase separation mode.
  • Supports a broad pH range of 1 to 12.
  • Withstands maximum pressures up to 18000 psi (1240 Bar).
  • Utilizes spherical particles with 1.7 μm particle size.
  • Designed with a 130 Å pore size.
  • Includes 4 units per package.
Catalog No. 50-791-066 Supplier Waters Corp Supplier No. 176001889
Add to Cart
Add to Cart
Request a Quote
This method development kit is designed for low pH UPLC applications, incorporating advanced 1.7 μm BEH ShieldRP18 and Phenyl with 1.8 μm HSS C18 and C18 SB column chemistries. It facilitates various method development approaches, enabling efficient and effective analytical processes.
  • Accommodates diverse method development strategies.
  • Leverages UPLC technology for enhanced throughput and reduced analysis time.
  • Maintains separation integrity for reliable results.
  • Features reversed phase separation mode.
  • Supports a broad pH range of 1 to 12.
  • Withstands maximum pressures up to 18000 psi (1240 Bar).
  • Utilizes spherical particles with 1.7 μm particle size.
  • Designed with a 130 Å pore size.
  • Includes 4 units per package.