Learn More
Thermo Scientific Low-Pressure Solvent Line Kit
Supplier:  Thermo Scientific LC230
Encompass
The Low-Pressure Solvent Line Kit (P/N LC230) contains the Tees, check valves, and low-pressure solvent lines to connect one pump (A or B) to the waste system and to its corresponding solvent bottle and six-port rotary valve.
Catalog No. NC1819932