Learn More
Perkin Elmer US LLC Low Pressure Piston Backup Guide Bushing
Catalog No. 502117198
Encompass_Preferred
All PerkinElmer pumps contain a high and low pressure piston seal. Whenever replacing the low pressure seal, the Low Pressure Seal Back-Up Ring should also be replaced.
Catalog No. 50-211-7198 Supplier Perkin Elmer US LLC Supplier No. 02542313
All PerkinElmer pumps contain a high and low pressure piston seal. Whenever replacing the low pressure seal, the Low Pressure Seal Back-Up Ring should also be replaced.