Learn More
Perkin Elmer US LLC Low Pressure Piston Seal for Series 200 Micropump
Catalog No. 502113099
Encompass_Preferred
All PerkinElmer pumps contain a high and low pressure piston seal. Whenever replacing the low pressure seal, the Low Pressure Seal Back-Up Ring should also be replaced.
Catalog No. 50-211-3099 Supplier Perkin Elmer US LLC Supplier No. 09923366
All PerkinElmer pumps contain a high and low pressure piston seal. Whenever replacing the low pressure seal, the Low Pressure Seal Back-Up Ring should also be replaced.