Learn More
Braintree Scientific Low Pressure Regulator with Flowmeter (0.5-12 LPM)
SDP
Catalog No. 501954751
Encompass
Low Pressure Regulator with Flowmeter0.5-12 lpm AVMA compliant this regulator includes a six foot hose with quick-disconnect fitting that attaches to the stainless steel lids. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4751 Supplier Braintree Scientific Supplier No. EP1305
Low Pressure Regulator with Flowmeter0.5-12 lpm AVMA compliant this regulator includes a six foot hose with quick-disconnect fitting that attaches to the stainless steel lids. **Sold for research purposes only and not for human use**