Learn More
V&P Scientific Inc REAGENT RESERVOIR, Low Profile, 384 Well Format, Pyramid Bottom, Natural Polypropylene, 25/pack
Catalog No. NC0731766
Encompass
REAGENT RESERVOIR, Low Profile, 384 Well Format, Pyramid Bottom, .007mL Working Well Capacity, 19mm Height, 12mm Reservoir Depth, 92mL Max Capacity, SLAS Footprint, Natural Polypropylene
Catalog No. NC0731766 Supplier V&P Scientific Inc Supplier No. VP 574L
REAGENT RESERVOIR, Low Profile, 384 Well Format, Pyramid Bottom, .007mL Working Well Capacity, 19mm Height, 12mm Reservoir Depth, 92mL Max Capacity, SLAS Footprint, Natural Polypropylene