Learn More
Bio Rad Laboratories LOW-PROFILE 0.2 ML 8-TUBE STRI
Supplier:  Bio Rad Laboratories TLS0801
Encompass
Strip, Tube; Bio-Rad; 8-tube strips; Low profile; Without caps; Natural; 120 strips; PCR volumes of 5-125uL recommended; 0.2mL
Catalog No. 50-901-0031
Edge