Learn More
Analytical Sales & Services Low Profile, 12 Well Reagent Reservoir
Catalog No. 5000806451
Encompass
Low Profile, 12 Well Reagent Reservoir\n• 15mL/Well\n• 180mL Plate Volume\n• Size: 127.8 x 85.5 x 31.4\n• Clear Polypropylene\n• Certified DNase/RNase and Pyrogen Free\n\nQty/Pkg: 50
Catalog No. 50-008-06451 Supplier Analytical Sales & Services Supplier No. 21512
Low Profile, 12 Well Reagent Reservoir\n• 15mL/Well\n• 180mL Plate Volume\n• Size: 127.8 x 85.5 x 31.4\n• Clear Polypropylene\n• Certified DNase/RNase and Pyrogen Free\n\nQty/Pkg: 50