Learn More
Analytical Sales & Services Low Profile, 384 Well Reagent Reservoir
Catalog No. 5000804468
Encompass
Low Profile, 384 Well Reagent Reservoir\n• 384-Position Base\n• 185mL Total Volume\n• Size:  127.8 x 85.5 x 31.4\n• Pyramid Bottoms\n• Clear Polypropylene\n• Certified DNase/RNase and Pyrogen Free\n\nQty/Pkg: 50
Catalog No. 50-008-04468 Supplier Analytical Sales & Services Supplier No. 38488
Low Profile, 384 Well Reagent Reservoir\n• 384-Position Base\n• 185mL Total Volume\n• Size:  127.8 x 85.5 x 31.4\n• Pyramid Bottoms\n• Clear Polypropylene\n• Certified DNase/RNase and Pyrogen Free\n\nQty/Pkg: 50