Learn More
Analytical Sales & Services  Low Profile, 96 Well Reagent Reservoir
Catalog No. 5000806352
Encompass
 Low Profile, 96 Well Reagent Reservoir\n• 96 Well Format\n• 195mL Total Volume\n• Size: 127.8 x 85.5 x 31.4\n• Pyramid Bottoms\n• Clear Polypropylene\n• Certified DNase/RNase and Pyrogen Fre\n\nQty/Pkg: 50
Catalog No. 50-008-06352 Supplier Analytical Sales & Services Supplier No. 96073
Add to Cart
Add to Cart
 Low Profile, 96 Well Reagent Reservoir\n• 96 Well Format\n• 195mL Total Volume\n• Size: 127.8 x 85.5 x 31.4\n• Pyramid Bottoms\n• Clear Polypropylene\n• Certified DNase/RNase and Pyrogen Fre\n\nQty/Pkg: 50