Learn More
Leica Biosystems Low-profile disp.blades DB80LS
Catalog No. 502552454
Encompass_Preferred
Cut the thin sections needed for diagnostic clarity.The Leica DB80LS disposable blade offers a new level of quality with low-profile shape, a premium finish and edge-to-edge, blade-to-blade consistency.The premium rounded corners provide a more comfortable and safer user experience when handling the microtome blade.For use with Leica Biosystems microtomes and cryostats classified and labeled as in vitro diagnostic medical devices for histological medical diagnosis, e.g. cancer diagnosis.For in Vitro Diagnostic Use
This item is not returnable. View return policy
Catalog No. 50-255-2454 Supplier Leica Biosystems Supplier No. 14035843489
Cut the thin sections needed for diagnostic clarity.The Leica DB80LS disposable blade offers a new level of quality with low-profile shape, a premium finish and edge-to-edge, blade-to-blade consistency.The premium rounded corners provide a more comfortable and safer user experience when handling the microtome blade.For use with Leica Biosystems microtomes and cryostats classified and labeled as in vitro diagnostic medical devices for histological medical diagnosis, e.g. cancer diagnosis.For in Vitro Diagnostic Use