Learn More
Leica Biosystems Low-profile disp.blades DB80LX
Supplier:  Leica Biosystems 14035843497
Encompass_Preferred
Cut the thin sections needed for diagnostic clarity.The Leica DB80LS disposable blade offers a new level of quality with low-profile shape, a premium finish and edge-to-edge, blade-to-blade consistency.The premium rounded corners provide a more comfortable and safer user experience when handling the microtome blade.For use with Leica Biosystems microtomes and cryostats classified and labeled as in vitro diagnostic medical devices for histological medical diagnosis, e.g. cancer diagnosis.For in Vitro Diagnostic Use
Catalog No. 50-255-2452