Learn More
Bellco Glass, Inc. Low Profile IOB Assembly , 3L
SDP
Catalog No. 501944747
Encompass_Preferred
IOB assembly uses smooth bearing action and fits under the 100mm screw cap with ptfe cap seal and silicone gasket. Allows for smoother and better mixing inside the spinner flasks. MADE IN THE USA Dimensions: 3 x 5 1/4"
Catalog No. 50-194-4747 Supplier Bellco Glass, Inc. Supplier No. 196403420
IOB assembly uses smooth bearing action and fits under the 100mm screw cap with ptfe cap seal and silicone gasket. Allows for smoother and better mixing inside the spinner flasks. MADE IN THE USA Dimensions: 3 x 5 1/4"