Learn More
Mitegen Low Profile Lid For Use With Nanosoft Grid Boxes
SDP
Catalog No. 5000494996
Encompass
The Low Profile Lid is made of anodized aluminum and is screwed on and off of the grid box with a hex key/allen wrench.  The lid is compatiable with Nanosoft pin lid type grid boxes. The low thickness of the lid means you can stack 2 grid boxes and lids in 1 pocket of a cryoEM grid storage puck, doubling your storage capacity. The anodized aluminum is highly robust, and is less susceptible to static electricity than plastic lids. Recommended hex key for use with this lid is SKU M-NS-LPL-AW
Catalog No. 50-004-94996 Supplier Mitegen Supplier No. MNSLPL1
The Low Profile Lid is made of anodized aluminum and is screwed on and off of the grid box with a hex key/allen wrench.  The lid is compatiable with Nanosoft pin lid type grid boxes. The low thickness of the lid means you can stack 2 grid boxes and lids in 1 pocket of a cryoEM grid storage puck, doubling your storage capacity. The anodized aluminum is highly robust, and is less susceptible to static electricity than plastic lids. Recommended hex key for use with this lid is SKU M-NS-LPL-AW