Learn More
V&P Scientific Inc MAGNETIC RACK, 8 Strip PCR Tubes, 0.2mL Volume
Catalog No. NC1412683
Encompass
MAGNETIC BEAD SEPARATION RACK, 8 Strip PCR Tubes, 0.2mL Volume, 1 Sided Bead Pellet Location, Variable Positions, 52MGO, NdFeB Magnet, 9mm Center Spacing, 6mm Hole Diameter, HDPE (High Density Polyethylene)
Catalog No. NC1412683 Supplier V&P Scientific Inc Supplier No. VP 772F4
Add to Cart
Add to Cart
MAGNETIC BEAD SEPARATION RACK, 8 Strip PCR Tubes, 0.2mL Volume, 1 Sided Bead Pellet Location, Variable Positions, 52MGO, NdFeB Magnet, 9mm Center Spacing, 6mm Hole Diameter, HDPE (High Density Polyethylene)