Learn More
V&P Scientific Inc MAGNETIC RACK, Low Profile, 8 Strip PCR Tubes, 0.2mL Volume
Catalog No. NC0988547
Encompass
MAGNETIC BEAD SEPARATION RACK, Low Profile, 8 Strip PCR Tubes, 0.2mL Volume, 1 Sided Bead Pellet Location, Low Position, 52MGO, NdFeB Magnet, 9mm Center Spacing, 6mm Hole Diameter, HDPE (High Density Polyethylene)
Catalog No. NC0988547 Supplier V&P Scientific Inc Supplier No. VP 772F41
MAGNETIC BEAD SEPARATION RACK, Low Profile, 8 Strip PCR Tubes, 0.2mL Volume, 1 Sided Bead Pellet Location, Low Position, 52MGO, NdFeB Magnet, 9mm Center Spacing, 6mm Hole Diameter, HDPE (High Density Polyethylene)