Learn More
JDM LAB SOLUTIONS LLC LOW PROFILE MAGNETIC STIRRER
Catalog No. NC1721745
Encompass
Sleek low profile design with high grade of waterproofing. Speed 15 - 1500 rpm, capacity 500 ml
Catalog No. NC1721745 Supplier JDM LAB SOLUTIONS LLC Supplier No. MS01FUA
Sleek low profile design with high grade of waterproofing. Speed 15 - 1500 rpm, capacity 500 ml