Learn More
Greiner Bio-One Low Profile Polystyrene Microplate Lids
GREENER_CHOICE
Catalog No. 07000291 Shop All Greiner Bio-One Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
07-000-291 Sterile
1 options
Catalog No. 07-000-291 Supplier Greiner Bio-One Supplier No. 656191
Add to Cart
Add to Cart
For use with 384 well standard microplates. Greiner Bio-one Low Profile Polystyrene Microplate Lids is free of detectable DNase, RNase, human DNA.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Offer protection against contamination and evaporation during sample storage and cell cultivation
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
No. of Wells 384
Height (Metric) 6 mm
Condensation Rings No
No. per Case 200
Plate Compatibility 384-well Standard Microplate
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.