Learn More
Analytical Sales & Services Low Profile Reservoir, 16 Channel - IRRADIATED
Catalog No. 5000807015
Encompass
Clear Polypropylene Reservoir 16 Channel Low Profile 4.9mL/Channel, 78mL Total Volume, Pyramid Bottoms - IRRADIATED\n\nQty/Pkg: 25
Catalog No. 50-008-07015 Supplier Analytical Sales & Services Supplier No. 32019
Clear Polypropylene Reservoir 16 Channel Low Profile 4.9mL/Channel, 78mL Total Volume, Pyramid Bottoms - IRRADIATED\n\nQty/Pkg: 25