Learn More
Analytical Sales & Services Low Profile Reservoir, Open Flat Bottom
Catalog No. 5000804287
Encompass
Low Profile, Single Cavity Reagent Reservoir\n• Open Flat Bottom\n• 190mL Total Volume\n• Size: 127.8 x 85.5 x 31.4\n• Clear Polypropylene\n• Certified DNase/RNase and Pyrogen Free\n\nQty/Pkg: 50
Catalog No. 50-008-04287 Supplier Analytical Sales & Services Supplier No. 38190
Low Profile, Single Cavity Reagent Reservoir\n• Open Flat Bottom\n• 190mL Total Volume\n• Size: 127.8 x 85.5 x 31.4\n• Clear Polypropylene\n• Certified DNase/RNase and Pyrogen Free\n\nQty/Pkg: 50